2-Chlorbenzylamin 96%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
(2-Chlorophenyl)methanamine
A15594.14
A15594.14EA
40.9
EUR
A15594.14
2-Chlorbenzylamin 96%
2-Chlorbenzylamin
Purity: 96%
CAS No.: 89-97-4
Molecular Formula: C7H8ClN
Formula Weight: 141.60
Flash Point: 88°(190°F)
Refractive Index: 1.5620
Odour: Amine-like
Boiling Point: 227°
Beilstein Reference: 907186
UN No.: 2735
MDL No.: MFCD00008108
Formel:
C₇H₈ClN Molecular Weight: 141,6 g/mol Siedepunkt: 227 °C Dichte: 1,170 Flammpunkt: 88 °C (190 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008108 CAS-Nummer: 89-97-4 EINECS: 201-955-8 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...