4-Fluoranilin ≥99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
A15597.14
A15597.22
A15597.36
A15597.14EA
22.5
EUR
A15597.14
A15597.22
A15597.36
4-Fluoranilin ≥99%
4-Fluoranilin
Purity: 99%
CAS No.: 371-40-4
Molecular Formula: C6H6FN
Formula Weight: 111.12
Flash Point: 73°(163°F)
Refractive Index: 1.5400
Odour: Recognizable
Melting Point: -2°
Boiling Point: 186-187°
Beilstein Reference: 742030
Merck Reference: 14,4169
UN No.: 2941
MDL No.: MFCD00007829
Formel:
C₆H₆FN Siedepunkt: 186…187 °C Schmelzpunkt: –2 °C Dichte: 1,158 Flammpunkt: 73 °C (163 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007829 CAS-Nummer: 371-40-4 EINECS: 206-735-5 UN: 2941 ADR: 6.1,III Merck Index: 14,4169 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...