3,4-Difluorbenzonitril ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
A15600.06
A15600.14
A15600.22
A15600.06EA
42.1
EUR
A15600.06
A15600.14
A15600.22
3,4-Difluorbenzonitril ≥98%
3,4-Difluorbenzonitril
Purity: 98%
CAS No.: 64248-62-0
Molecular Formula: C7H3F2N
Formula Weight: 139.11
Flash Point: 69°(156°F)
Melting Point: 48-52°
Boiling Point: 180°
Beilstein Reference: 2082224
UN No.: 3439
MDL No.: MFCD00011666
Formel:
C₇H₃F₂N Siedepunkt: 180 °C Schmelzpunkt: 48…52 °C Flammpunkt: 69 °C (156 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011666 CAS-Nummer: 64248-62-0 EINECS: 264-751-8 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...