4-tert-Butylcyclohexanon ≥99%
Lieferant: Thermo Fisher Scientific
A15612.22
A15612.22EA
56.1
EUR
A15612.22
4-tert-Butylcyclohexanon ≥99%
4-tert-Butylcyclohexanon
Purity: 99%
CAS No.: 98-53-3
Molecular Formula: C10H18O
Formula Weight: 154.25
Flash Point: 96°(204°F)
Melting Point: 47-51°
Boiling Point: 113-116°/20mm
Beilstein Reference: 507309
MDL No.: MFCD00001642
Formel:
C₁₀H₁₈O Siedepunkt: 113…116 °C (20 mmHg) Schmelzpunkt: 47…51 °C Dichte: 0,893 Flammpunkt: 96 °C (204 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001642 CAS-Nummer: 98-53-3 EINECS: 202-678-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...