3-Bromanisol ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A15625.14
A15625.22
A15625.36
A15625.14EA
38.5
EUR
A15625.14
A15625.22
A15625.36
3-Bromanisol ≥98%
3-Bromanisol
Purity: 98+%
CAS No.: 2398-37-0
Molecular Formula: C7H7BrO
Formula Weight: 187.04
Flash Point: 93°(199°F)
Refractive Index: 1.5640
Melting Point: ca 2°
Boiling Point: 210-211°
Beilstein Reference: 1524687
MDL No.: MFCD00000081
Formel:
C₇H₇BrO Siedepunkt: 210…211 °C Schmelzpunkt: ∼2 °C Dichte: 1,489 Flammpunkt: 93 °C (199 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000081 CAS-Nummer: 2398-37-0 EINECS: 219-264-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...