4-Brombutylacetat ≥98%

Lieferant: Thermo Fisher Scientific

Synonyme: Essigsäure(4-brombutyl)ester

A15632.09 A15632.18 A15632.30
A15632.09EA 45.8 EUR
A15632.09 A15632.18 A15632.30
4-Brombutylacetat ≥98%
4-Brombutylacetat

Purity: 98+% CAS No.: 4753-59-7 Molecular Formula: CH3CO2(CH2)4Br Formula Weight: 195.07 Flash Point: 109°(228°F) Refractive Index: 1.4600 Boiling Point: 96-97°/15mm Beilstein Reference: 1749695 MDL No.: MFCD00000263

Formel: C₆H₁₁BrO₂
Siedepunkt: 96…97 °C (15 mmHg)
Dichte: 1,348
Flammpunkt: 109 °C (228 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00000263
CAS-Nummer: 4753-59-7
EINECS: 225-277-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR