(3-Fluorphenyl)essigsäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-(3-Fluorophenyl)acetic acid

A15635.06 A15635.14 A15635.22
A15635.06EA 49.6 EUR
A15635.06 A15635.14 A15635.22
(3-Fluorphenyl)essigsäure ≥98%
(3-Fluorphenyl)essigsäure

Purity: 98% CAS No.: 331-25-9 Molecular Formula: C8H7FO2 Formula Weight: 154.14 Flash Point: 110°(230°F) Odour: Odourless Boiling Point: 151°/0.8mm Beilstein Reference: 775898 MDL No.: MFCD00004331

Formel: (C₆H₄F)CH₂COOH
Molecular Weight: 154.14 g/mol
Siedepunkt: 151 °C (0.8 mmHg)
Schmelzpunkt: 44…48 °C
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004331
CAS-Nummer: 331-25-9
EINECS: 206-360-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR