(Methyl)triphenylphosphoniumiodid ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

A15644.14 A15644.22 A15644.36
A15644.14EA 58.5 EUR
A15644.14 A15644.22 A15644.36
(Methyl)triphenylphosphoniumiodid ≥98%
(Methyl)triphenylphosphoniumiodid

Purity: 98% CAS No.: 2065-66-9 Molecular Formula: CH3P(C6H5)3I Formula Weight: 404.23 Odour: Odourless Melting Point: 185-188° UN No.: 3464 MDL No.: MFCD00066175

Formel: C₁₉H₁₈IP
Schmelzpunkt: 185…188 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00066175
CAS-Nummer: 2065-66-9
EINECS: 218-178-5
UN: 3464
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR