(3,4-Dichlorphenyl)essigsäure ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3,4-Dichlorophenylacetic acid
A15653.06
A15653.14
A15653.22
A15653.06EA
25.2
EUR
A15653.06
A15653.14
A15653.22
(3,4-Dichlorphenyl)essigsäure ≥98%
(3,4-Dichlorphenyl)essigsäure
Purity: 98%
CAS No.: 5807-30-7
Molecular Formula: C8H6Cl2O2
Formula Weight: 205.04
Odour: Odourless
Melting Point: 87-92°
Beilstein Reference: 1953781
MDL No.: MFCD00004333
Formel:
(C₆H₃Cl₂)CH₂COOH Molecular Weight: 205.04 g/mol Siedepunkt: 314 Schmelzpunkt: 87…92 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004333 CAS-Nummer: 5807-30-7 EINECS: 227-368-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...