Methyldecanoat ≥99%

Lieferant: Thermo Fisher Scientific

A15658.18 A15658.22 A15658.30
A15658.18EA 43.5 EUR
A15658.18 A15658.22 A15658.30
Methyldecanoat ≥99%
Methyldecanoat

Purity: 99% CAS No.: 110-42-9 Molecular Formula: CH3(CH2)8CO2CH3 Formula Weight: 186.30 Flash Point: 94°(201°F) Refractive Index: 1.4250 Melting Point: -18° Boiling Point: 223-225° Beilstein Reference: 1759170 MDL No.: MFCD00009580

Formel: C₁₁H₂₂O₂
Siedepunkt: 223…225 °C
Schmelzpunkt: –18 °C
Dichte: 0,877
Flammpunkt: 94 °C (201 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00009580
CAS-Nummer: 110-42-9
EINECS: 203-766-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR