Dicyclohexylamin 98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: DCHA

A15671.0F A15671.AE A15671.AP
A15671.0FEA 92 EUR
A15671.0F A15671.AE A15671.AP
Dicyclohexylamin 98%
Dicyclohexylamin

Purity: 98% CAS No.: 101-83-7 Molecular Formula: C12H23N Formula Weight: 181.32 Flash Point: 99°(210°F) Refractive Index: 1.4842 Melting Point: -2° Boiling Point: 256° Beilstein Reference: 605923 Merck Reference: 14,3095 UN No.: 2565 MDL No.: MFCD00011658

Formel: C₁₂H₂₃N
Molecular Weight: 181.32 g/mol
Siedepunkt: 256 °C
Schmelzpunkt: –2 °C
Dichte: 0.913
Flammpunkt: 99 °C (210 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011658
CAS-Nummer: 101-83-7
EINECS: 202-980-7
UN: 2565
ADR: 8 III
Merck Index: 13,03122

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR