2-Chlor-3,5-dinitrobenzoesäure, (max. 5% H₂O) 97% (Trockengewicht)

Lieferant: Thermo Fisher Scientific
Achtung

A15675.14 A15675.22
A15675.14EA 21 EUR
A15675.14 A15675.22
2-Chlor-3,5-dinitrobenzoesäure, (max. 5% H₂O) 97% (Trockengewicht)
2-Chloro-3,5-dinitrobenzoic acid

Purity: 97% CAS No.: 2497-91-8 Molecular Formula: C7H3ClN2O6 Formula Weight: 246.56 Melting Point: 196-201° Beilstein Reference: 2146480

Formel: C₇H₃ClN₂O₆
Molecular Weight: 246.56 g/mol
Schmelzpunkt: 196…201 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007070
CAS-Nummer: 2497-91-8
EINECS: 219-684-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR