Triethanolamin Hydrochlorid ≥99%
Lieferant: Thermo Fisher Scientific
A15678.0B
A15678.0I
A15678.30
A15678.0BEA
163
EUR
A15678.0B
A15678.0I
A15678.30
Triethanolamin Hydrochlorid ≥99%
Triethanolamin Hydrochlorid
Purity: 99+%
CAS No.: 637-39-8
Molecular Formula: C6H15NO3.HCl
Formula Weight: 185.65
Melting Point: 177-179°
Beilstein Reference: 3909940
Merck Reference: 14,9665
MDL No.: MFCD00012596
Formel:
C₆H₁₆ClNO₃ Schmelzpunkt: 177…179 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012596 CAS-Nummer: 637-39-8 EINECS: 211-284-2 Merck Index: 14,9665 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...