Pyridiniumtribromid tech. 90%, technische Qualität
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
A15684.14
A15684.22
A15684.36
A15684.14EA
43.6
EUR
A15684.14
A15684.22
A15684.36
Pyridiniumtribromid tech. 90%, technische Qualität
Pyridinium tribromide
Purity: 90%
CAS No.: 39416-48-3
Molecular Formula: C5H5N.HBr3
Formula Weight: 319.82
Melting Point: 128-136°
Beilstein Reference: 3690144
Merck Reference: 14,7973
UN No.: 3261
MDL No.: MFCD00013223
Formel:
C₅H₆Br₃N Schmelzpunkt: 128…136 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013223 CAS-Nummer: 39416-48-3 EINECS: 254-446-8 UN: 3261 ADR: 8,III Merck Index: 14,7973 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...