Methyl-o-toluat ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-Tolylsäuremethylester, o-Toluylsäuremethylester, 2-Methylbenzoesäuremethylester, Methyl-2-methylbenzoat, 2-Toluylsäuremethylester
A15695.14
A15695.22
A15695.36
A15695.14EA
33.2
EUR
A15695.14
A15695.22
A15695.36
Methyl-o-toluat ≥99%
Methyl-o-toluat
Purity: 99%
CAS No.: 89-71-4
Molecular Formula: C9H10O2
Formula Weight: 150.18
Flash Point: 82°(179°F)
Refractive Index: 1.5190
Boiling Point: 207-208°
Beilstein Reference: 2041846
MDL No.: MFCD00008428
Formel:
C₉H₁₀O₂ Molecular Weight: 150.18 Siedepunkt: 207…208 °C Schmelzpunkt: –50 °C Dichte: 1.072 Flammpunkt: 82 °C (179 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008428 CAS-Nummer: 89-71-4 EINECS: 201-932-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...