2-Chlorbenzamid ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 2-Chlorbenzoesäureamid

A15706.14 A15706.22
A15706.14EA 36.5 EUR
A15706.14 A15706.22
2-Chlorbenzamid ≥98%
2-Chlorbenzamid

Purity: 98% CAS No.: 609-66-5 Molecular Formula: C7H6ClNO Formula Weight: 155.58 Melting Point: 140-144° Beilstein Reference: 508510

Formel: C₇H₆ClNO
Molecular Weight: 155.58 g/mol
Schmelzpunkt: 140…144 °C
Dichte: 1.34 g/cm³ (18 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00007973
CAS-Nummer: 609-66-5
EINECS: 210-195-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR