Pyridinium-p-toluolsulfonat ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

A15708.14 A15708.22 A15708.36
A15708.14EA 84.3 EUR
A15708.14 A15708.22 A15708.36
Pyridinium-p-toluolsulfonat ≥98%
Pyridinium-p-toluolsulfonat

Purity: 98+% CAS No.: 24057-28-1 Molecular Formula: C12H13NO3S Formula Weight: 251.31 Melting Point: 116-120° Beilstein Reference: 3764305

Formel: C₁₂H₁₃NO₃S
Schmelzpunkt: 116…120 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00013108
CAS-Nummer: 24057-28-1
EINECS: 246-002-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR