Ethylencarbonat ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 1,3-Dioxa-2-cyclopentanon, Ethylenglycolcarbonat

A15735.0C A15735.0E A15735.36
A15735.0CEA 394 EUR
A15735.0C A15735.0E A15735.36
Ethylencarbonat ≥99%
Ethylencarbonat

Purity: 99% CAS No.: 96-49-1 Molecular Formula: C3H4O3 Formula Weight: 88.06 Flash Point: 160°(320°F) Melting Point: 35-37° Boiling Point: 244-245° Solubility: Miscible (40%) with water, alchohol ethyl acetate, benzene and chloroform. Soluble in ether, n-Butanol, and carbon tetrachloride. Beilstein Reference: 106249 MDL No.: MFCD00005382

Formel: C₃H₄O₃
Molecular Weight: 88.06 g/mol
Siedepunkt: 244…245 °C
Schmelzpunkt: 35…37 °C
Dichte: 1.320
Flammpunkt: 160 °C (320 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00005382
CAS-Nummer: 96-49-1
EINECS: 202-510-0

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR