1-Brom-4-chlor-3-fluorbenzol ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A15745.14
A15745.22
A15745.14EA
81.4
EUR
A15745.14
A15745.22
1-Brom-4-chlor-3-fluorbenzol ≥98%
1-Brom-4-chlor-3-fluorbenzol
Formel:
C₆H₃BrClF Siedepunkt: 62…64 °C (9 mmHg) Dichte: 1,710 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042519 CAS-Nummer: 60811-18-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...