3-Methylbenzo[b]thiophen ≥98%
Lieferant: Thermo Fisher Scientific
A15772.03
A15772.06
A15772.14
A15772.03EA
46.1
EUR
A15772.03
A15772.06
A15772.14
3-Methylbenzo[b]thiophen ≥98%
3-Methylbenzo[b]thiophen
Purity: 98%
CAS No.: 1455-18-1
Molecular Formula: C9H8S
Formula Weight: 148.23
Flash Point: 104°(219°F)
Refractive Index: 1.6240
Melting Point: -40°
Boiling Point: 72-74°/2mm
Beilstein Reference: 111282
MDL No.: MFCD00040243
Formel:
C₉H₈S Siedepunkt: 72…74 °C (2 mmHg) Schmelzpunkt: –40 °C Dichte: 1,13 Flammpunkt: 104 °C (219 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00040243 CAS-Nummer: 1455-18-1 EINECS: 215-934-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...