L-Alanin 99

Lieferant: Thermo Fisher Scientific

Synonyme: H-L-Ala-OH, (S)-(+)-2-Aminopropionic acid, L(+)-2-Aminopropionic acid, A, (S)-(+)-Alanin, α-Alanin, H-Ala-OH, (S)-(+)-2-Aminopropionsäure, L(+)-Alanin, (S)-(+)-2-Aminopropansäure, L-α-Alanin

A15804.14 A15804.22 A15804.36
A15804.14EA 30.3 EUR
A15804.14 A15804.22 A15804.36
L-Alanin 99
L-Alanin

Purity: 99% CAS No.: 56-41-7 Molecular Formula: C3H7NO2 Formula Weight: 89.09 Melting Point: ca 295° dec. Beilstein Reference: 1720248 Merck Reference: 14,204 Optical Rotation: +14° (c=5 in 5N HCl) MDL No.: MFCD00064410

Formel: C₃H₇NO₂
Molecular Weight: 89.09 g/mol
Siedepunkt: 189
Schmelzpunkt: ∼ 295 °C (Zers.)
Dichte: 1.432
Lagertemperatur: Raumtemperatur
MDL: MFCD00064410
CAS-Nummer: 56-41-7
EINECS: 200-273-8
Merck Index: 13,00203

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR