(±)-Epichlorhydrin ≥99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Epichlorhydrin, 1-Chlor-2,3-epoxypropan, 2,3-Epoxypropylchlorid, (±)-1-Chlor-2,3-epoxypropan, 3-Chlor-1,2-epoxypropan, 2-Chlormethyloxiran, (±)-Glycerolepichlorhydrin, (±)-3-Chlor-1,2-epoxypropan
Purity: 99%
CAS No.: 106-89-8
Molecular Formula: C3H5ClO
Formula Weight: 92.53
Flash Point: 32°(90°F)
Physical Form: Liquid
Density: 1.182
Refractive Index: 1.4380
Appearance: Colourless
Odour: Like chlorine
Melting Point: -57°
Boiling Point: 115-117°
Beilstein Reference: 79785
Merck Reference: 14,3611
UN No.: 2023
MDL No.: MFCD00005132
A cyclodextrin cross-linking reagent
Formel:
C₃H₅ClO Siedepunkt: 115…117 °C Schmelzpunkt: –57 °C Dichte: 1,182 Flammpunkt: 32 °C (90 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005132 CAS-Nummer: 106-89-8 EINECS: 203-439-8 UN: 2023 ADR: 6.1,II Merck Index: 14,3611 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...