Tetraphenylphosphoniumbromid ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

A15860.09 A15860.18 A15860.22
A15860.09EA 37.4 EUR
A15860.09 A15860.18 A15860.22
Tetraphenylphosphoniumbromid ≥98%
Tetraphenylphosphonium bromide

Purity: 98+% CAS No.: 2751-90-8 Molecular Formula: (C6H5)4PBr Formula Weight: 419.31 Melting Point: 296-302° Beilstein Reference: 3922383 Merck Reference: 14,9237 MDL No.: MFCD00011915

Formel: C₂₄H₂₀BrP
Schmelzpunkt: 296…302 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00011915
CAS-Nummer: 2751-90-8
EINECS: 220-393-4
Merck Index: 14,9237

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR