(3,4-Dihydroxyphenyl)essigsäure ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
DOPAC, 3,4-Dihydroxybenzeneacetic acid, 3,4-Dihydroxyphenylacetic acid
A15893.03
A15893.06
A15893.14
A15893.03EA
34.7
EUR
A15893.03
A15893.06
A15893.14
(3,4-Dihydroxyphenyl)essigsäure ≥98%
(3,4-Dihydroxyphenyl)essigsäure
Purity: 98+%
CAS No.: 102-32-9
Molecular Formula: C8H8O4
Formula Weight: 168.15
Melting Point: 128-130°
Beilstein Reference: 2211017
Formel:
(C₆H₃(OH)₂)CH₂COOH Molecular Weight: 168.15 g/mol Siedepunkt: 337 Schmelzpunkt: 128…130 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004338 CAS-Nummer: 102-32-9 EINECS: 203-024-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...