α,α,α-Trifluor-m-toluidin ≥99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
3-Aminobenzotrifluorid, 3-(Trifluormethyl)anilin
A15910.0E
A15910.22
A15910.36
A15910.0EEA
302
EUR
A15910.0E
A15910.22
A15910.36
α,α,α-Trifluor-m-toluidin ≥99%
α,α,α-Trifluor-m-toluidin
Purity: 99%
CAS No.: 98-16-8
Molecular Formula: C7H6F3N
Formula Weight: 161.13
Flash Point: 85°(185°F)
Refractive Index: 1.4800
Odour: Recognizable
Melting Point: 5-6°
Boiling Point: 187-189°
Beilstein Reference: 387672
UN No.: 2948
MDL No.: MFCD00007797
Formel:
C₇H₆F₃N Molecular Weight: 161.13 g/mol Siedepunkt: 187…189 °C Schmelzpunkt: 5…6 °C Dichte: 1.295 Flammpunkt: 85 °C (185 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007797 CAS-Nummer: 98-16-8 EINECS: 202-643-4 UN: 2948 ADR: 6.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...