1-Brom-4-hexylbenzol ≥97%

Lieferant: Thermo Fisher Scientific

Synonyme: p-Brom-hexylbenzol

A15918.06 A15918.14
A15918.06EA 93 EUR
A15918.06 A15918.14
1-Brom-4-hexylbenzol ≥97%
1-Brom-4-hexylbenzol

Purity: 97% CAS No.: 23703-22-2 Molecular Formula: C12H17Br Formula Weight: 241.17 Flash Point: 120°(248°F) Refractive Index: 1.5213 Boiling Point: 280° MDL No.: MFCD00061114

Formel: C₁₂H₁₇Br
Molecular Weight: 241.17 g/mol
Siedepunkt: 280 °C
Dichte: 1.179
Flammpunkt: >120 °C (248 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00061114
CAS-Nummer: 23703-22-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR