Diacetone acrylamide 99%

Lieferant: Thermo Fisher Scientific
Achtung

A15940.0B A15940.18 A15940.30
A15940.0BEA 336 EUR
A15940.0B A15940.18 A15940.30
Diacetone acrylamide 99%
Diacetone acrylamide

Purity: 99% CAS No.: 2873-97-4 Molecular Formula: CH3COCH2C(CH3)2NHCOCH=CH2 Formula Weight: 169.23 Flash Point: 110°(230°F) Melting Point: 54-56° Beilstein Reference: 1928444 MDL No.: MFCD00008788

Formel: C₉H₁₅NO₂
Schmelzpunkt: 54…56 °C
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008788
CAS-Nummer: 2873-97-4
EINECS: 220-713-2
Merck Index: 14,2963

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR