4-Brom-α,α,α-trifluortoluol ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A15942.06
A15942.14
A15942.22
A15942.06EA
45.6
EUR
A15942.06
A15942.14
A15942.22
4-Brom-α,α,α-trifluortoluol ≥99%
4-Brom-α,α,α-trifluortoluol
Purity: 99%
CAS No.: 402-43-7
Molecular Formula: C7H4BrF3
Formula Weight: 225.01
Flash Point: 60°(140°F)
Refractive Index: 1.4730
Boiling Point: 154-155°
Beilstein Reference: 2045666
UN No.: 1993
MDL No.: MFCD00000398
Formel:
C₇H₄BrF₃ Siedepunkt: 154…155 °C Dichte: 1,630 Flammpunkt: 60 °C (140 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000398 CAS-Nummer: 402-43-7 EINECS: 206-943-6 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...