trans-2-Octen-1-ol ≥97%
Lieferant: Thermo Fisher Scientific
Achtung
|
A15960.06
A15960.14
A15960.22
A15960.06EA
47.4
EUR
A15960.06
A15960.14
A15960.22
trans-2-Octen-1-ol ≥97%
trans-2-Octen-1-ol
Purity: 97%
CAS No.: 18409-17-1
Molecular Formula: C8H16O
Formula Weight: 128.22
Flash Point: 85°(185°F)
Refractive Index: 1.4465
Boiling Point: 88°/11mm
Beilstein Reference: 1720707
MDL No.: MFCD00014057
Formel:
C₈H₁₆O Siedepunkt: 88 °C (11 mmHg) Dichte: 0,850 Flammpunkt: 85 °C (185 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014057 CAS-Nummer: 18409-17-1 EINECS: 242-288-2 UN: 1987 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...