Nicotinamid ≥99%
Lieferant: Thermo Fisher Scientific
Purity: 99%
CAS No.: 98-92-0
Molecular Formula: C6H6N2O
Formula Weight: 122.13
Flash Point: 182°(360°F)
Physical Form: Powder
Density: 1.40
Appearance: White
Odour: Odourless
Melting Point: 128-132°
Beilstein Reference: 383619
Merck Reference: 14,6523
MDL No.: MFCD00006395
The amide of nicotinic acid (vitamin B-3)
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...