1,3,5-Trifluorbenzol 98
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
A15984.06
A15984.06EA
67.5
EUR
A15984.06
1,3,5-Trifluorbenzol 98
1,3,5-Trifluorobenzene
Purity: 98+%
CAS No.: 372-38-3
Molecular Formula: C6H3F3
Formula Weight: 132.09
Flash Point: -7°(19°F)
Refractive Index: 1.4150
Melting Point: -7 to -4°
Boiling Point: 75-76°
Beilstein Reference: 1906457
UN No.: 2387
MDL No.: MFCD00000333
Formel:
C₆H₃F₃ Molecular Weight: 132.09 g/mol Siedepunkt: 75…76 °C Schmelzpunkt: –7 …–4 °C Dichte: 1.279 Flammpunkt: –7 °C (19 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000333 CAS-Nummer: 372-38-3 EINECS: 206-751-2 UN: 2387 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...