4,7-Dichlorchinolin ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

A15985.14 A15985.22 A15985.36
A15985.14EA 62.4 EUR
A15985.14 A15985.22 A15985.36
4,7-Dichlorchinolin ≥98%
4,7-Dichloroquinoline

Purity: 98% CAS No.: 86-98-6 Molecular Formula: C9H5Cl2N Formula Weight: 198.05 Flash Point: 164°(327°F) Melting Point: 83-87° Boiling Point: 148°/10mm Beilstein Reference: 125359 MDL No.: MFCD00006774

Formel: C₉H₅Cl₂N
Molecular Weight: 198.05 g/mol
Siedepunkt: 148 °C (10 mmHg)
Schmelzpunkt: 83…87 °C
Flammpunkt: 164 °C (327 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00006774
CAS-Nummer: 86-98-6
EINECS: 201-714-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR