4,5-Dibrom-3(2H)-pyridazinon ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4,5-Dibromopyridazin-3(2H)-one

A15994.06 A15994.14
A15994.06EA 91.2 EUR
A15994.06 A15994.14
4,5-Dibrom-3(2H)-pyridazinon ≥98%
4,5-Dibrom-3(2H)-pyridazinon

Purity: 98% CAS No.: 5788-58-9 Molecular Formula: C4H2Br2N2O Formula Weight: 253.89 Melting Point: ca 220° dec. Beilstein Reference: 121469

Formel: C₄H₂Br₂N₂O
Molecular Weight: 253.89 g/mol
Schmelzpunkt: ∼ 220 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00023641
CAS-Nummer: 5788-58-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR