Theophyllin Monohydrat ≥99%

Lieferant: Thermo Fisher Scientific
Gefahr

A15997.30 A15997.18 A15997.0B
A15997.30EA 75.5 EUR
A15997.30 A15997.18 A15997.0B
Theophyllin Monohydrat ≥99%

Purity: 99% CAS No.: 5967-84-0 Molecular Formula: C7H8N4O2.H2O Formula Weight: 198.18 (180.16anhy) Odour: Odourless Melting Point: 270-274° Beilstein Reference: 13463 Merck Reference: 14,9285 UN No.: 2811 MDL No.: MFCD00151659

Formel: C₇H₈N₄O₂·1H₂O
Schmelzpunkt: 270…274 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00151659
CAS-Nummer: 5967-84-0
EINECS: 200-385-7
UN: 2811
ADR: 6.1,III
Merck Index: 14,9285

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR