N-Butylphthalimid ≥99%
Lieferant: Thermo Fisher Scientific
Synonyme:
2-Butylisoindoline-1,3-dione, N-(n-Butyl)phthalimide
A16020.06
A16020.14
A16020.06EA
28.5
EUR
A16020.06
A16020.14
N-Butylphthalimid ≥99%
N-Butylphthalimid
Purity: 99%
CAS No.: 1515-72-6
Molecular Formula: C12H13NO2
Formula Weight: 203.24
Melting Point: 32-35°
Boiling Point: 120-123°/5mm
MDL No.: MFCD00039695
Formel:
C₁₂H₁₃NO₂ Siedepunkt: 120…123 °C (5 mmHg) Schmelzpunkt: 32…35 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00039695 CAS-Nummer: 1515-72-6 EINECS: 216-157-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...