2'-Deoxyuridin ≥99%
Lieferant: Thermo Fisher Scientific
A16026.03
A16026.06
A16026.14
A16026.03EA
22.5
EUR
A16026.03
A16026.06
A16026.14
2'-Deoxyuridin ≥99%
2'-Deoxyuridin
Purity: 99%
CAS No.: 951-78-0
Molecular Formula: C9H12N2O5
Formula Weight: 228.21
Melting Point: 164-167°
Beilstein Reference: 24433
Merck Reference: 14,2910
Optical Rotation: +52° (c=1.1 in 1N NaOH)
MDL No.: MFCD00006527
Formel:
C₉H₁₂N₂O₅ Schmelzpunkt: 164…167 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006527 CAS-Nummer: 951-78-0 EINECS: 213-455-7 Merck Index: 14,2910 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...