1,4-Phenylendiessigsäure ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2,2'-(1,4-Phenylene)diacetic acid

A16036.03 A16036.06
A16036.03EA 24.1 EUR
A16036.03 A16036.06
1,4-Phenylendiessigsäure ≥97%
1,4-Phenylendiessigsäure

Purity: 97% CAS No.: 7325-46-4 Molecular Formula: C10H10O4 Formula Weight: 194.19 Odour: Odourless Melting Point: 252-256° Beilstein Reference: 1960342 MDL No.: MFCD00004354

Formel: C₁₀H₁₀O₄
Schmelzpunkt: 252…256 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004354
CAS-Nummer: 7325-46-4
EINECS: 230-801-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR