Indigocarmin
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Acid Blue 74, Food blue No. 2
A16052.14
A16052.22
A16052.14EA
29.2
EUR
A16052.14
A16052.22
Indigocarmin
Indigocarmin
CAS No.: 860-22-0
Molecular Formula: C16H8N2Na2O8S2
Formula Weight: 466.36
Odour: Odourless
Melting Point:
Formel:
C₁₆H₈N₂Na₂O₈S₂ Schmelzpunkt: >300 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005723 CAS-Nummer: 860-22-0 EINECS: 212-728-8 Merck Index: 14,4944 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...