Indigocarmin

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Acid Blue 74, Food blue No. 2

A16052.14 A16052.22
A16052.14EA 29.2 EUR
A16052.14 A16052.22
Indigocarmin
Indigocarmin

CAS No.: 860-22-0 Molecular Formula: C16H8N2Na2O8S2 Formula Weight: 466.36 Odour: Odourless Melting Point: 300° Beilstein Reference: 4103904 Merck Reference: 14,4944 MDL No.: MFCD00005723

Formel: C₁₆H₈N₂Na₂O₈S₂
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005723
CAS-Nummer: 860-22-0
EINECS: 212-728-8
Merck Index: 14,4944

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR