Cyclohexan ≥99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 99+%
CAS No.: 110-82-7
Molecular Formula: C6H12
Formula Weight: 84.16
Flash Point: -18°(-1°F)
Refractive Index: 1.4260
Odour: Pungent
Melting Point: 6.5°
Boiling Point: 80-81°
Beilstein Reference: 1900225
Merck Reference: 14,2723
UN No.: 1145
MDL No.: MFCD00003814
Formel:
C₆H₁₂ Siedepunkt: 80…81 °C Schmelzpunkt: 6,5 °C Dichte: 0,779 Flammpunkt: –18 °C (–1 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003814 CAS-Nummer: 110-82-7 EINECS: 203-806-2 UN: 1145 ADR: 3,II Merck Index: 14,2723 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...