5-Brom-2-fluorbenzonitril ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

A16113.14 A16113.06
A16113.14EA 89.6 EUR
A16113.14 A16113.06
5-Brom-2-fluorbenzonitril ≥98%
5-Brom-2-fluorbenzonitril

Purity: 98% CAS No.: 179897-89-3 Molecular Formula: C7H3BrFN Formula Weight: 200.01 Melting Point: 77-81° Beilstein Reference: 7701063 UN No.: 3439 MDL No.: MFCD00143424

Formel: C₇H₃BrFN
Siedepunkt: 255 °C (1013 hPa)
Schmelzpunkt: 77…81 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00143424
CAS-Nummer: 179897-89-3
UN: 3439
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR