Cobalt(II,III)oxid

Lieferant: Thermo Fisher Scientific
Achtung

A16121.0B A16121.18 A16121.30
A16121.0BEA 355 EUR
A16121.0B A16121.18 A16121.30
Cobalt(II,III)oxid
Cobalt(II,III)oxid

CAS No.: 1308-06-1 Molecular Formula: Co3O4 Formula Weight: 240.80 Melting Point: 900° dec. Merck Reference: 14,2429 UN No.: 3077 MDL No.: MFCD00010939

Formel: Co₃O₄
Dichte: 6,11
Lagertemperatur: Raumtemperatur
MDL: MFCD00010939
CAS-Nummer: 1308-06-1
EINECS: 215-157-2
ADR: 9,III
Merck Index: 14,2429

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR