L(+)-Lysin Dihydrochlorid ≥99%
Lieferant: Thermo Fisher Scientific
Synonyme:
H-Lys-OH.2HCl, (S)-(+)-2,6-Diaminohexansäure Dihydrochlorid, (S)-(+)-2,6-Diaminocapronsäure Dihydrochlorid, (S)-(+)-Lysin Dihydrochlorid
A16127.14
A16127.22
A16127.14EA
35
EUR
A16127.14
A16127.22
L(+)-Lysin Dihydrochlorid ≥99%
L(+)-Lysin Dihydrochlorid
Purity: 99%
CAS No.: 657-26-1
Molecular Formula: C6H14N2O2.2HCl
Formula Weight: 219.11
Melting Point: 202-206°
Beilstein Reference: 4539715
Merck Reference: 14,5636
MDL No.: MFCD00066389
Formel:
C₆H₁₆Cl₂N₂O₂ Molecular Weight: 219.11 g/mol Schmelzpunkt: 202…206 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00066389 CAS-Nummer: 657-26-1 EINECS: 211-518-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...