Quecksilber(II)iodid ≥99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
A16130.22
A16130.36
A16130.22EA
101
EUR
A16130.22
A16130.36
Quecksilber(II)iodid ≥99%
Purity: 99+%
CAS No.: 7774-29-0
Molecular Formula: HgI2
Formula Weight: 454.40
Odour: Odourless
Melting Point: 259°
Boiling Point: 350° subl.
Merck Reference: 14,5879
UN No.: 1638
MDL No.: MFCD00011044
Formel:
HgI₂ Siedepunkt: 350 °C (Subl.) Schmelzpunkt: 259 °C Dichte: 6,28 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011044 CAS-Nummer: 7774-29-0 EINECS: 231-873-8 UN: 1638 ADR: 6.1,II Merck Index: 14,5879 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...