Benzethoniumchlorid ≥97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: Hyamine® 1622

A16142.22
A16142.22EA 50.3 EUR
A16142.22
Benzethoniumchlorid ≥97%
Benzethoniumchlorid

Purity: 97% CAS No.: 121-54-0 Molecular Formula: C27H42ClNO2 Formula Weight: 448.10 Odour: Odourless Melting Point: 157-165° Beilstein Reference: 3898548 Merck Reference: 14,1074 UN No.: 1759 MDL No.: MFCD00011742

Formel: C₂₇H₄₂ClNO₂
Molecular Weight: 448,09 g/mol
Schmelzpunkt: 157…165 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00011742
CAS-Nummer: 121-54-0
EINECS: 204-479-9
UN: 1759
ADR: 8,III
Merck Index: 14,1074

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR