Propionaldehyd ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Propanal
A16146.0J
A16146.AP
A16146.AU
A16146.0JEA
162
EUR
A16146.0J
A16146.AP
A16146.AU
Propionaldehyd ≥97%
Propionaldehyd
Purity: 97%
CAS No.: 123-38-6
Molecular Formula: CH3CH2CHO
Formula Weight: 58.08
Storage Temperature: 2°C to 8°C
Flash Point: -40°(-40°F)
Refractive Index: 1.3620
Odour: Pungent
Melting Point: -81°
Boiling Point: 47-49°
Beilstein Reference: 506010
Merck Reference: 14,7823
UN No.: 1275
MDL No.: MFCD00007020
Formel:
C₃H₆O Siedepunkt: 47…49 °C Schmelzpunkt: –81 °C Dichte: 0,81 Flammpunkt: –40 °C (–40 °F) Lagertemperatur: Kühlschrank |
MDL:
MFCD00007020 CAS-Nummer: 123-38-6 EINECS: 204-623-0 UN: 1275 ADR: 3,II Merck Index: 14,7823 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...