Quecksilber(II)oxid, rot 99%

Lieferant: Thermo Fisher Scientific
Gefahr

A16157.0B A16157.18 A16157.30
A16157.0BEA 322 EUR
A16157.0B A16157.18 A16157.30
Quecksilber(II)oxid, rot 99%
Mercury (II) oxide

Purity: 99% CAS No.: 21908-53-2 Molecular Formula: HgO Formula Weight: 216.59 Odour: Odourless Melting Point: ca 500° dec. UN No.: 1641 MDL No.: MFCD00011045

Formel: HgO
Siedepunkt: 356,6 °C (1013 hPa)
Dichte: 11,14
Lagertemperatur: Raumtemperatur
MDL: MFCD00011045
CAS-Nummer: 21908-53-2
EINECS: 244-654-7
UN: 1641
ADR: 6.1,II
Merck Index: 14,5882

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR