Iodmonobromid ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

A16158.14 A16158.22 A16158.30
A16158.14EA 32.6 EUR
A16158.14 A16158.22 A16158.30
Iodmonobromid ≥98%
Iodmonobromid

Purity: 98% CAS No.: 7789-33-5 Molecular Formula: IBr Formula Weight: 206.81 Storage Temperature: 2°C to 8°C Flash Point: None Odour: Pungent Melting Point: 50° subl. Boiling Point: 116° dec. Beilstein Reference: 3902973 Merck Reference: 14,5016 UN No.: 3260 MDL No.: MFCD00011353

Formel: IBr
Siedepunkt: 116 °C (Zersetzung)
Schmelzpunkt: 50 °C (Subl.)
Dichte: 4,416
Lagertemperatur: Kühlschrank
MDL: MFCD00011353
CAS-Nummer: 7789-33-5
EINECS: 232-159-9
UN: 3260
ADR: 8,II
Merck Index: 14,5016

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR