Kaliumiodat ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
A16162.0E
A16162.22
A16162.36
A16162.0EEA
541
EUR
A16162.0E
A16162.22
A16162.36
Kaliumiodat ≥98%
Kaliumiodat
Purity: 98%
CAS No.: 7758-05-6
Molecular Formula: KIO3
Formula Weight: 214.00
Melting Point: 560° dec.
Merck Reference: 14,7642
MDL No.: MFCD00011406
Formel:
KlO₃ Schmelzpunkt: 560 °C (Zers.) Dichte: 3,89 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011406 CAS-Nummer: 7758-05-6 EINECS: 231-831-9 UN: 1479 ADR: 5.1,II Merck Index: 14,7642 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...