L(+)-Rhamnose Monohydrat ≥99%

Lieferant: Thermo Fisher Scientific

Synonyme: Isodulcit, 6-Desoxy-L(+)-mannose Monohydrat

A16166.06 A16166.14 A16166.22
A16166.06EA 40.1 EUR
A16166.06 A16166.14 A16166.22
L(+)-Rhamnose Monohydrat ≥99%
L(+)-Rhamnose Monohydrat

Purity: 99% CAS No.: 10030-85-0 Molecular Formula: C6H12O5.H2O Formula Weight: 182.19 (164.17anhy) Melting Point: 89-94° Beilstein Reference: 5988592 Merck Reference: 14,8172 Optical Rotation: +8° (c=10 in water, 20h) MDL No.: MFCD00149363

Formel: C₆H₁₂O₅·1H₂O
Molecular Weight: 182.17 g/mol
Schmelzpunkt: 89…94 °C
Dichte: 1.47
Lagertemperatur: Raumtemperatur
MDL: MFCD00149363
CAS-Nummer: 10030-85-0
EINECS: 222-793-4
Merck Index: 12,08338

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR